Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSESSKKRK PKVIRSDGAP AEGKRNRSDT EQEGKYYSEE AEVDLRDPGR DYELYKYTCQ ELQRLMAEIQ DLKSRGGKDV AIEIEERRIQ SCVHFMTLKK LNRLAHIRLK KGRDQTHEAK QKVDAYHLQL QNLLYEVMHL QKEITKCLEF KSKHEEIDLV SLEEFYKEAP PDISKAEVTM GDPHQQTLAR LDWELEQRKR LAEKYRECLS NKEKILKEIE VKKEYLSSLQ PRLNSIMQAS LPVQEYLFMP FDQAHKQYET ARHLPPPLYV LFVQATAYGQ ACDKTLSVAI EGSVDEAKAL FKPPEDSQDD ESDSDAEEEQ TTKRRRPTLG VQLDDKRKEM LKRHPLSVML DLKCKDDSVL HLTFYYLMNL NIMTVKAKVT TAMELITPIS AGDLLSPDSV LSCLYPGDHG KKTPNPANQY QFDKVGILTL SDYVLELGHP YLWVQKLGGL HFPKEQPQQT VIADHSLSAS HMETTMKLLK TRVQSRLALH KQFASLEHGI VPVTSDCQYL FPAKVVSRLV KWVTIAHEDY MELHFTKDIV DAGLAGDTNL YYMALIERGT AKLQAAVVLN PGYSSIPPIF QLCLNWKGEK TNSNDDNIRA MEGEVNVCYK ELCGPWPSHQ LLTNQLQRLC VLLDVYLETE SHDDSVEGPK EFPQEKMCLR LFRGPSRMKP FKYNHPQGFF SHR
Component the THO subcomplex of the TREX complex. The TREX complex specifically associates with spliced mRNA and not with unspliced pre-mRNA. It is recruited to spliced mRNAs by a transcription-independent mechanism. Binds to mRNA upstream of the exon-junction complex (EJC) and is recruited in a splicing- and cap-dependent manner to a region near the 5' end of the mRNA where it functions in mRNA export. The recruitment occurs via an interaction between THOC4 and the cap-binding protein NCBP1. The TREX complex is essential for the export of Kaposi's sarcoma-associated herpesvirus (KSHV) intronless mRNAs and infectious virus production. The recruitment of the TREX complex to the intronless viral mRNA occurs via an interaction beteen KSHV ORF57 protein and THOC4. UAP56 functions as a bridge between THOC4 and the THO complex. THOC5 in conjunction with THOC4 functions in NXF1-NXT1 mediated nuclear export of HSP70 mRNA. May be involved in the differentiation of granulocytes and adipocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Fms-interacting protein; fSAP79; Functional spliceosome-associated protein 79; hTREX90; NF2/meningioma region protein pK1.3; Placental protein 39.2; PP39.2; THO complex subunit 5
Gene Aliases: C22orf19; Fmip; fSAP79; KIAA0983; PK1.3; THOC5
UniProt ID: (Human) Q13769
Entrez Gene ID: (Human) 8563
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support