Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MDDADPEER NYDNMLKMLS DLNKDLEKLL EEMEKISVQA TWMAYDMVVM RTNPTLAESM RRLEDAFVNC KEEMEKNWQE LLHETKQRL (1-88 aa encoded by BC127859)
Major component of the transverse central element of synaptonemal complexes (SCS), formed between homologous chromosomes during meiotic prophase. Required for chromosome loading of the central element-specific SCS proteins, and for initiating synapsis between homologous chromosomes. Chromosome loading appears to require SYCP1. Required for fertility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptonemal complex central element protein 3; Testis highly expressed gene 2 protein; testis highly expressed protein 2; Testis-specific expressed protein 2; THEG-2; TSEG-2
Gene Aliases: 1700007E06Rik; C22orf41; SYCE3; THEG2; TSEG-2; TSEG2
UniProt ID: (Human) A1L190, (Mouse) B5KM66
Entrez Gene ID: (Human) 644186, (Mouse) 75459
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support