Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: LMAFQKYSGE TVQERKQKDR KALHELKLEE WKGRLQVTEH LPEKIESSLQ EDEPENDAKK IEALLNLPRN PSVIDKQDKD; Positive Samples: THP-1, U-937, A-375; Cellular Location: Mitochondrion membrane, Multi-pass membrane protein
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Complex I assembly factor TIMMDC1, mitochondrial; M5-14 protein; Protein M5-14; TIMM domain containing-protein 1; Translocase of inner mitochondrial membrane domain-containing protein 1; transmembrane protein C3orf1; transmembrane protein C3orf1 homolog
Gene Aliases: 2810021C21Rik; 4930455C21Rik; AV135763; C3orf1; RGD1359380; TIMMDC1; UNQ247/PRO284
UniProt ID: (Human) Q9NPL8, (Mouse) Q8BUY5, (Rat) Q6AY94
Entrez Gene ID: (Human) 51300, (Mouse) 76916, (Rat) 303922
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support