Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MKEALHQIV VRCSELVSST SLPRLSVSRL QGPPDSQPLG TLGQGGWKLL GIVGSLAPET LGGLGTEFGP CTHPLPFDMV RERERDDELR QGWLLQCPQC ARTLLCHCGP FLTPPSQTSS SGFQLCSLKP SGSLVTATEP LSNFAFSYFP (1-149 aa encoded by BC146513)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: C3orf27; GR6
Entrez Gene ID: (Human) 23434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support