Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKEALHQIVV RCSELVSSTS LPRLSVSRLQ GPPDSQPLGT LGQGGWKLLG IVGSLAPETL GGLGTEFGPC THPLPFDMVR ERERDDELRQ GWLLQCPQCA RTLLCHCGPF LTPPSQTSSS GFQLCSLKPS GSLVTATEPL SNFAFSYFP
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene Aliases: C3orf27; GR6
Entrez Gene ID: (Human) 23434
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support