Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
DIPK2A (CC058, 1190002N15Rik) may play a role in cardiomyocyte proliferation through paracrine signaling and activation of the PPI3K-AKT-CDK7 signaling cascade
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cc058; deleted in autism 1; Deleted in autism protein 1; Deleted in autism protein 1 homolog; Divergent protein kinase domain 2A; Golgi protein GoPro49; Golgi Protein of 49 kDa; GoPro49; HASF; hypoxia and Akt induced stem cell factor; Hypoxia and AKT-induced stem cell factor; UPF0672 protein C3orf58; UPF0672 protein C3orf58 homolog
Gene Aliases: Ab2-095; C3orf58; DIA1; DIPK2A; GoPro49; HASF
UniProt ID: (Mouse) Q3USZ8, (Human) Q8NDZ4
Entrez Gene ID: (Mouse) 68861, (Human) 205428, (Rat) 315891
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support