Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Human: 93%
SETD9 gene ontology annotations related to this gene include regulation of signal transduction by p53 class mediator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SET domain-containing protein 9
Gene Aliases: C5orf35; SETD9
UniProt ID: (Human) Q8NE22
Entrez Gene ID: (Human) 133383
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support