Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: HRDDREHAKS LGLHVNEEEQ ERPVGVLTSS VYGKRINQPI EPLNRDFGRA NHVQADFYRK NDIPSLKEPG FGHIAPS
Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 83%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 90
Gene Aliases: C5orf49; CFAP90
UniProt ID: (Human) A4QMS7
Entrez Gene ID: (Human) 134121
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support