Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
PXDC1 gene ontology annotations related to this gene include phosphatidylinositol binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PX domain-containing protein 1; PX domain-containing protein C6orf145; PX domain-containing protein C6orf145 homolog
Gene Aliases: 1300014I06Rik; C6orf145; PXDC1; RGD1311307
UniProt ID: (Human) Q5TGL8, (Mouse) Q8JZU6, (Rat) Q4KLK8
Entrez Gene ID: (Human) 221749, (Mouse) 66895, (Rat) 361238
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support