Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MTPESRDTTD LSPGGTQEME GIVIVKVEEE DEEDHFQKER NKVESSPQVL SRSTTMNERA LLSSYLVAYR VAKEKMAHTA AEKIILPACM DMVRTIFDDK SADKLRTIPL SDNTISRRIC TIAKHLEAML ITRLQSGIDF AIQLDESTDI ASCPTLLVYV RYVWQDDFVE DLLCCLNLNS HITGLDLFTE LENCLLGQYK LNWKHCKGIS SDGTANMTGK HSRLTEKLLE ATHNNAVWNH CFIHREALVS KEISPSLMDV LKNAVKTVNF IKGSSLNSRL LEIFCSEIGV NHTHLLFHTE VRWLSQGKVL SRVYELRNEI YIFLVEKQSH LANIFEDDIW VTKLAYLSDI FGILNELSLK MQGKNNDIFQ YLEHILGFQK TLLLWQARLK SNRPSYYMFP TLLQHIEENI INEDCLKEIK LEILLHLTSL SQTFNYYFPE EKFESLKENI WMKDPFAFQN PESIIELNLE PEEENELLQL SSSFTLKNYY KILSLSAFWI KIKDDFPLLS RKSILLLLPF TTTYLCELGF SILTRLKTKK RNRLNSAPDM RVALSSCVPD WKELMNRQAH PSH
This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 200, member A; Protein FAM200A
Gene Aliases: C7orf38; FAM200A
UniProt ID: (Human) Q8TCP9
Entrez Gene ID: (Human) 221786
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support