Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMSTEQMQPL EVSEDRLDKL DPRCSHLDDL SDQFIKDCDL KKKPRKGKNV QATLNVESDQ KKPRRKDTPA LHIPPFIPGV FSEHLIKRYD VQERHPKGKM IPVLHNTDLE QKKPRRKDTP ALHMSPFAAG VTLLRDERPK AIVEDDEKDG DKIAI
Inhibits phosphatase activities of protein phosphatase 1 (PP1) and protein phosphatase 2A (PP2A) complexes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-substrate; Protein phosphatase 1 regulatory subunit 17; protein phosphatase 1, regulatory subunit 17
Gene Aliases: C7orf16; GSBS; PPP1R17
UniProt ID: (Human) O96001
Entrez Gene ID: (Human) 10842
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support