Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LLSFFIYNPR FGPREGQEEN KILFYHPNEV EKNEKIRNVG LCEAIVQFTR TFSPSKPAKS LHTQKNRQFF NEPEENFWMV MVVRNPIIEK QSKDGKPVIE Y
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Acts in concert with MON1A, as a guanine exchange factor (GEF) for RAB7, promotes the exchange of GDP to GTP, converting it from an inactive GDP-bound form into an active GTP-bound form (PubMed:23084991). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CCZ1 vacuolar protein trafficking and biogenesis associated homolog; H_NH0577018.2; Vacuolar fusion protein CCZ1 homolog; Vacuolar fusion protein CCZ1 homolog B; Vacuolar fusion protein CCZ1 homolog-like
Gene Aliases: C7orf28A; C7orf28B; CCZ1; CCZ1A; CCZ1B; CGI-43; H_DJ1163J12.2
UniProt ID: (Human) P86790
Entrez Gene ID: (Human) 51622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support