Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
RBM48 gene ontology annotations related to this gene include RNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA-binding protein 48; UPF0712 protein C7orf64; UPF0712 protein C7orf64 homolog
Gene Aliases: AW548102; C030048B08Rik; C7orf64; HSPC304; MGC16142; RBM48; RGD1310794
UniProt ID: (Human) Q5RL73, (Rat) Q561R3, (Mouse) Q8K2X2
Entrez Gene ID: (Human) 84060, (Rat) 296840, (Mouse) 269623
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support