Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAALGHLAGE AAAAPGPGTP CASRGARLPG PVSSARNPST VCLCPEQPTC SNADSRAHPL GDEGGTASKK QKNKKKTRNR ASVANGGEKA SEKLAPEEVP LSAEAQAQQL AQELAWCVEQ LELGLKRQKP TPKQKEQAIG AIRTLRSKRT PLPRKRQLMH SLFGDYRAQM EAEWREALRA LRAAAYSAQV QPVDGATRKK SQRVCRPRSI WRAKATLDMP DEEFRFNFF
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPF0488 protein C8orf33
Gene Aliases: C8orf33
UniProt ID: (Human) Q9H7E9
Entrez Gene ID: (Human) 65265
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support