Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
MCMDC2 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MCM domain-containing protein 2; MCM domain-containing protein C8orf45; MCM domain-containing protein C8orf45 homolog; Minichromosome maintenance domain-containing protein 2
Gene Aliases: 6030422M02Rik; C8orf45; MCMDC2
UniProt ID: (Human) Q4G0Z9, (Mouse) E9Q956, (Rat) Q5XI14
Entrez Gene ID: (Human) 157777, (Mouse) 240697, (Rat) 500392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support