Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene plays a role in the nonhomologous end joining (NHEJ) pathway of DNA double-strand break repair. The encoded protein may function to stabilize the Ku70/Ku80 heterodimer to facilitate the assembly and maintain the stability of the NHEJ complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Paralog of XRCC4 and XLF; Protein PAXX; uncharacterized protein C9orf142 homolog; XRCC4-like small protein
Gene Aliases: C9orf142; D930050G13Rik; PAXX; XLS
UniProt ID: (Mouse) Q8K0Y7, (Human) Q9BUH6
Entrez Gene ID: (Mouse) 227622, (Human) 286257, (Rat) 296565
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support