Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein has not been specifically defined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Behcet's Disease Associated Gene 1; coiled-coil domain containing 180; Coiled-coil domain-containing protein 180
Gene Aliases: BDAG1; C9orf174; CCDC180; Gm1025; KIAA1529
UniProt ID: (Human) Q9P1Z9
Entrez Gene ID: (Human) 100499483, (Rat) 100910460, (Mouse) 381522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support