Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDLPDESQWD ETTCGLAVCQ HPQCWATIRR IERGHPRILG SSCKTPLDAE DKLPVLTVVD ILDSGFAAHH LPECTFTKAH SLLSQSSKFY SKFHGRPPKG LPDKSLINCT NRLPKFPVLN LNETQLPCPE DVRNMVVLWI PEETEIHVSQ HGKKKRKNSA VKSKSFLGLS GNQSAGTRVG TPGMIVPPPT PVQLSEQFSS DFLPLWAQSE ALPQDLLKEL LPGGKQTMLC PEMKIKLAMM KKNLPLEKNR PDSVISSKMF LSIHRLTLER PALRYPERLK KLHNLKTEGY RKQQQRQQQQ QQQQKKVKTP IKKQEAKKKA KSDPGIQSTS HKHPVTTVHD RLYGYRTLPG QNSDMKQQQQ MEKGTTSKQD STERPKMNYY DHADFHHSVK SPELYETEPT NKDISAPVDA VPEAQAARQK KISFNFSEIM ASTGWNSELK LLRILQDTDD EDEEDQSSGA E
C9orf43 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Uncharacterized protein C9orf43
Gene Aliases: C9orf43
UniProt ID: (Human) Q8TAL5
Entrez Gene ID: (Human) 257169
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support