Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALLGSRAEL EADEDVFEDA LETISISSHS DMATSSLHFA SCDTQQAPRQ RGASTVSSSS STKVDLKSGL EECAVALNLF LSNKFTDALE LLRPWAKESM YHALGYSTIV VLQAVLTFEQ QDIQNGISAM KDALQTCQKY RKKYTVVESF SSLLSRGSLE QLSEEEMHAE ICYAECLLQK AALTFVQDEN MINFIKGGLK IRTSYQIYKE CLSILHEIQK NKLQQEFFYE FEGGVKLGSG AFNLMLSLLP ARIIRLLEFI GFSGNRELGL LQLREGASGR SMRSALCCLT ILAFHTYISL ILGTGEVNVA EAERLLAPFL QQFPNGSLVL FYHARIELLK GNLEEAQEVF QKCVSVQEEW KQFHHLCYWE LMWINVFQQN WMQAYYYSDL LCKESKWSKA TYVFLKAAIL SMLPEEDVVA TNENVVTLFR QVDSLKQRIA GKSIPTEKFA VRKARRYSAS LPAPVKLILP ALEMMYVWNG FSIVSKRKDL SENLLVTVEK AEAALQSQNF NSFSVDDECL VKLLKGCCLK NLQRPLQAEL CYNHVVESEK LLKYDHYLVP FTLFELASLY KSQGEIDKAI KFLETARNNY KDYSLESRLH FRIQAALHLW RKPSSD
Regulates high density lipoprotein (HDL) cholesterol metabolism by promoting the ubiquitination and degradation of the oxysterols receptors LXR (NR1H2 and NR1H3). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tetratricopeptide repeat protein 39B; TPR repeat protein 39B
Gene Aliases: C9orf52; TTC39B
UniProt ID: (Human) Q5VTQ0
Entrez Gene ID: (Human) 158219
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support