Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSSSGGAPGA SASSAPPAQE EGMTWWYRWL CRLSGVLGAV SCAISGLFNC ITIHPLNIAA GVWMIMNAFI LLLCEAPFCC QFIEFANTVA EKVDRLRSWQ KAVFYCGMAV VPIVISLTLT TLLGNAIAFA TGVLYGLSAL GKKGDAISYA RIQQQRQQAD EEKLAETLEG EL
Transmembrane protein which mediates synaptic endocytosis and fitness-based cell culling. In response to different stimulus strengths, controls two major modes of synaptic vesicle (SV) retrieval in hippocampal neurons; Clathrin- mediated endocytosis (CME) in response to mild stimulation and activity-dependent bulk endocytosis (ADBE) in response to strong stimulation (By similarity). In cytotoxic T-lymphoocytes (CTLs) facilitates calcium-dependent endocytosis of cytotoxic granules at the immuno synapse (By similarity). Different isoforms work as fitness fingerprints in 'loser' and 'winner' cells and thereby mediate win/lose decisions as part of the cell competition process.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium channel flower domain-containing protein 1; Calcium channel flower homolog; likely ortholog of H. sapiens chromosome 9 open reading frame 7 (C9orf7)
Gene Aliases: 5930434B04Rik; C9orf7; CACFD1; D9S2135; FLOWER; mFwe; PSEC0107; PSEC0248; RGD1311501; UNQ3071/PRO9903
UniProt ID: (Human) Q9UGQ2, (Rat) D4A9I3, (Mouse) Q8BG21
Entrez Gene ID: (Human) 11094, (Rat) 296599, (Mouse) 381356
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support