Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MISSKPRLVV PYGLKTLLEG ISRAVLKTNP SNINQFAAAY FQELTMYRGN TTMDIKDLVK QFHQIKVEKW SEGTTPQKKL ECLKEPGKTS VESKVPTQME KSTDTDEDNV TRTEYSDKTT QFPSVYAVPG TEQTEAVGGL SSKPATPKTT TPPSSPPPTA VSPEFAYVPA DPAQLAAQML AMATSERGQP PPCSNMWTLY CLTDKNQQGH PSPPPAPGPF PQATLYLPNP KDPQFQQHPP KVTFPTYVMG DTKKTSAPPF ILVGSNVQEA QGWKPLPGHA VVSQSDVLRY VAMQVPIAVP ADEKYQKHTL SPQNANPPSG QDVPRPKSPV FLSVAFPVED VAKKSSGSGD KCAPFGSYGI AGEVTVTTAH KRRKAETEN
Spermatozoa gain fertilization capacitation and hyperactivation after residing in the uterus or oviduct. Calcium, cAMP and protein tyrosine phosphorylation are necessary for the molecular mechanisms that allow for this transformation. CABYR (Calcium-binding tyrosine phosphorylation-regulated protein), also known as Fibrousheathin-2 or Cancer/testis antigen 88, is a 493 amino acid protein that is expressed in sperm flagella and exhibits increased tyrosine phosphorylation during capacitation. There are six named isoforms of CABYR that are produced as a result of alternative slicing events. Specifically, isoform 1 is expressed in the testis, while isoform 3 and isoform 5 are expressed in pancreas, brain and various brain tumors. Isoforms 1, 2 and 6 most likely bind calcium, whereas isoforms 3, 4 and 5 probably do not bind calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); Calcium-binding protein 86; Calcium-binding tyrosine phosphorylation-regulated protein; calcium-binding tyrosine-(Y)-phosphorylation regulated (fibrousheathin 2); Cancer/testis antigen 88; CT88; fibrousheathin 2; Fibrousheathin II; Fibrousheathin-2; FSP-2; testis tissue sperm-binding protein Li 84P; Testis-specific calcium-binding protein CBP86
Gene Aliases: CABYR; CABYRa; CABYRc; CABYRc/d; CABYRe; CBP86; CT88; FSP-2; FSP2
UniProt ID: (Human) O75952
Entrez Gene ID: (Human) 26256
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support