Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CaV2.1 Antibodies

          Osenses

          CACNA1A Polyclonal Antibody

          View all (12) CaV2.1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CACNA1A Polyclonal Antibody

          Product Details

          OSC00271W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:2,000
          -

          Immunohistochemistry (IHC)

          1:300-1:2,000
          -
          Product Specifications

          Species Reactivity

          Mouse, Rat

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from aa region of 2150-2210 of rat CACNA1A conjugated to blue carrier protein was used as the antigen
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The peptide is homologous in mouse.

          Specificity of this antibody: CACNA1A.

          Target Information

          Voltage-dependent calcium channels mediate the entry of calcium ions into excitable cells, and are also involved in a variety of calcium-dependent processes, including muscle contraction, hormone or neurotransmitter release, and gene expression. Calcium channels are multisubunit complexes composed of alpha-1, beta, alpha-2/delta, and gamma subunits. The channel activity is directed by the pore-forming alpha-1 subunit, whereas, the others act as auxiliary subunits regulating this activity. The distinctive properties of the calcium channel types are related primarily to the expression of a variety of alpha-1 isoforms, alpha-1A, B, C, D, E, and S. This gene encodes the alpha-1A subunit, which is predominantly expressed in neuronal tissue. Mutations in this gene are associated with 2 neurologic disorders, familial hemiplegic migraine and episodic ataxia 2. This gene also exhibits polymorphic variation due to n-repeats. Multiple transcript variants encoding different isoforms have been found for this gene. In one set of transcript variants, the n-repeats occur in the 3 UTR, and are not associated with any disease. But in another set of variants, an insertion extends the coding region to include the n-repeats which encode a polyglutamine tract. Expansion of the n-repeats from the normal 4-16 to 21-28 in the coding region is associated with spinocerebellar ataxia 6.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BI; brain calcium channel 1; Brain calcium channel I; brain class A; calcium channel alpha 1A; calcium channel, L type, alpha-1 polypeptide; Calcium channel, L type, alpha-1 polypeptide isoform 4; Calcium channel, L type, alpha-1 polypeptide, isoform 4; calcium channel, voltage-dependent, alpha 1A subunit; leaner; Rat brain class A; RBA-I; rocker; tottering; Voltage-dependent P/Q-type calcium channel subunit alpha-1A; Voltage-gated calcium channel subunit alpha Cav2.1

          View more View less

          Gene Aliases: alpha1A; APCA; BccA1; BI; Caca1a; Cach4; Cacn3; Cacna1a; Cacnl1a4; Cav2.1; Ccha1a; EA2; FHM; HPCA; la; MHP; MHP1; nmf352; rbA-1; rkr; SCA6; tg

          View more View less

          UniProt ID: (Mouse) P97445, (Rat) P54282

          View more View less

          Entrez Gene ID: (Mouse) 12286, (Rat) 25398

          View more View less

          Function(s)
          DNA binding ion channel activity voltage-gated ion channel activity voltage-gated calcium channel activity calcium channel activity protein binding calmodulin binding high voltage-gated calcium channel activity syntaxin binding metal ion binding
          Process(es)
          sulfur amino acid metabolic process glucose metabolic process transport ion transport calcium ion transport positive regulation of cytosolic calcium ion concentration gamma-aminobutyric acid signaling pathway chemical synaptic transmission neuron-neuron synaptic transmission neuromuscular synaptic transmission synapse assembly adult walking behavior cell death gamma-aminobutyric acid secretion regulation of acetylcholine secretion, neurotransmission cell growth calcium ion regulated exocytosis regulation of calcium ion-dependent exocytosis transmission of nerve impulse sensory perception of pain spinal cord motor neuron differentiation cerebellum maturation cerebellar molecular layer development cerebellar Purkinje cell layer development cerebellar Purkinje cell differentiation vestibular nucleus development central nervous system neuron differentiation cellular chloride ion homeostasis negative regulation of hormone biosynthetic process regulation of ion transmembrane transport synaptic transmission, glutamatergic neurotransmitter metabolic process regulation of membrane potential thyroid hormone metabolic process hormone metabolic process receptor clustering negative regulation of neuron apoptotic process response to pain behavioral response to pain calcium ion-regulated exocytosis of neurotransmitter dendrite morphogenesis regulation of axonogenesis neurological system process musculoskeletal movement, spinal reflex action neuromuscular process controlling balance neuromuscular process membrane depolarization synaptic transmission, GABAergic transmembrane transport rhythmic synaptic transmission calcium ion import calcium ion transmembrane transport membrane depolarization during action potential
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0