Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Predicted to react with Mouse and Rat samples.
L-type calcium channels are composed of five subunits. The protein Calcium Channel 5 represents one of these subunits, gamma, and is one of several gamma subunit proteins. It is an integral membrane protein that is thought to stabilize the calcium channel in an inactive (closed) state.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium channel, voltage-dependent, gamma subunit 5; Neuronal voltage-gated calcium channel gamma-5 subunit; TARP gamma-5; Transmembrane AMPAR regulatory protein gamma-5; voltage dependent calcium channel gamma 5 subunit; Voltage-dependent calcium channel gamma-5 subunit
Gene Aliases: CACNG5
UniProt ID: (Human) Q9UF02, (Rat) Q8VHW8, (Mouse) Q8VHW4
Entrez Gene ID: (Human) 27091, (Rat) 140726, (Mouse) 140723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support