Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: NATPQVAMQV LEIHYTPSVK IIPSTPFPQE GQPLILTCES KGKPLPEPVL WTKDGGELPD PDRMVVSGRE LNILFLNKTD NGTYRCEATN TIGQSSAEYV LIVHDPNALA GQNGPD; Positive Samples: Mouse liver; Cellular Location: Cell junction, Cell membrane, Cell projection, Single-pass type I membrane protein, axon, synapse
This gene encodes a member of the synaptic cell adhesion molecule 1 (SynCAM) family which belongs to the immunoglobulin (Ig) superfamily. The encoded protein has three Ig-like domains and a cytosolic protein 4.1 binding site near the C-terminus. Proteins belonging to the protein 4.1 family crosslink spectrin and interact with other cytoskeletal proteins. Multiple transcript variants encoding different isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell adhesion molecule 2; IgSF4D; Immunoglobulin superfamily member 4D; immunoglobulin superfamily, member 4; NECL-3; Nectin-like protein 3; Synaptic cell adhesion molecule 2; SynCAM 2
Gene Aliases: 2900078E11Rik; 9330131D06; A830029E02Rik; Cadm2; Igdf4d; Igsf4d; NECL3; SynCAM 2; SynCAM2
UniProt ID: (Mouse) Q8BLQ9
Entrez Gene ID: (Mouse) 239857
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support