Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CADM3 Antibodies

          Proteintech

          CADM3 Polyclonal Antibody

          View all (13) CADM3 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CADM3 Polyclonal Antibody

          • Antibody Testing Data (4)
          CADM3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CADM3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CADM3 Antibody (15660-1-AP) in WB

          Various lysates were subjected to SDS PAGE followed by western blot with (Product # 15660-1-AP) CADM3 antibody at dilution of 1:1,000 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CADM3 Antibody in Western Blot (WB)
          CADM3 Antibody in Western Blot (WB)
          CADM3 Antibody in Western Blot (WB)
          CADM3 Antibody in Flow Cytometry (Flow)

          Product Details

          15660-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CADM3 Fusion Protein Ag8081 (1-398 aa encoded by BC033819)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.33 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGAPAASLL LLLLLFACCW APGGANLSQD DSQPWTSDET VVAGGTVVLK CQVKDHEDSS LQWSNPAQQT LYFGEKRALR DNRIQLVTST PHELSISISN VALADEGEYT CSIFTMPVRT AKSLVTVLGI PQKPIITGYK SSLREKDTAT LNCQSSGSKP AARLTWRKGD QELHGEPTRI QEDPNGKTFT VSSSVTFQVT REDDGASIVC SVNHESLKGA DRSTSQRIEV LYTPTAMIRP DPPHPREGQK LLLHCEGRGN PVPQQYLWEK EGSVPPLKMT QESALIFPFL NKSDSGTYGC TATSNMGSYK AYYTLNVNDP SPVPSSSSTY HAIIGGIVAF IVFLLLIMLI FLGHYLIRHK GTYLTHEAKG SDDAPDADTA IINAEGGQSG GDDKKEYFI (1-398 aa encoded by BC033819 )

          Target Information

          IGSF4B is a brain-specific protein related to the calcium-independent cell-cell adhesion molecules known as nectins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Brain immunoglobulin receptor; Cell adhesion molecule 3; dendritic cell nectin-like protein 1 short isoform; IgSF4B; Immunoglobulin superfamily member 4B; immunoglobulin superfamily, member 4B; NECL-1; nectin-like 1; Nectin-like protein 1; nectin-lke 1; Synaptic cell adhesion molecule 3; SynCAM3; TSLC1-like 1; TSLC1-like protein 1; TSLL1

          View more View less

          Gene Aliases: BIgR; CADM3; IGSF4B; Necl-1; NECL1; SYNCAM3; TSLL1; UNQ225/PRO258

          View more View less

          UniProt ID: (Human) Q8N126, (Mouse) Q99N28, (Rat) Q1WIM3

          View more View less

          Entrez Gene ID: (Human) 57863, (Mouse) 94332, (Rat) 360882

          View more View less

          Function(s)
          receptor activity receptor binding protein homodimerization activity cell adhesion molecule binding protein binding
          Process(es)
          homophilic cell adhesion via plasma membrane adhesion molecules heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules cell recognition protein localization adherens junction organization cell adhesion
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0