Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EKPKRGSIG ENQIKDEKIK KDKEPKEEVK SFMDRKKGFT EVKSQNGEFM THKLKHTENT FSRPGGRASV DTKEAEGAPQ VEAGKRLEEL RRRRGETESE EFEKLKQKQQ EAALELEELK KKREERRKVL EEEEQRRKQE EADRKLREEE EKRRLKEEIE RRRAEAAEKR QKMPEDGLSD DKKPFKCFTP KGSSLKIEER AEFLNKSVQK SSGVKSTHQA AIVSKIDSRL EQYTSAIEGT KSAKPTKPAA SDLPVPAEGV RNIKSMWEKG NVFSSPTAAG TPNKETAGLK VGVSSRINEW LTKTPDGNKS PAPKPSDLRP GDVSSKRNLW EKQSVDKVTS PTKV (196-538 aa encoded by B C040354)
Caldesmon is a developmentally regulated protein involved in smooth muscle and non muscle contraction. Two closely related variants of human caldesmon have been identified which differ in their electrophorectic mobility and cellular distribution. The h caldesmon variant (120 - 150 kDa) is predominantly expressed in smooth muscle whereas l caldesmon (70 - 80 kDa) is found in non-muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Caldesmon; CDM; h-caldesmon; L-caldesmon; MGC21352; Non-muscle caldesmon; OTTHUMP00000207950; OTTHUMP00000207953; OTTHUMP00000207954; OTTHUMP00000207955; OTTHUMP00000214473; testis secretory sperm-binding protein Li 227n
Gene Aliases: 4833423D12Rik; AI195384; AV071549; AW536160; C920027I18Rik; CAD; CALD1; CDM; H-CAD; HCAD; L-CAD; LCAD; NAG22
UniProt ID: (Human) Q05682, (Rat) Q62736
Entrez Gene ID: (Human) 800, (Rat) 25687, (Mouse) 109624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support