Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Calmodulin 2 Antibodies

          Abnova

          CALM2 Polyclonal Antibody, MaxPab™

          View all (4) Calmodulin 2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CALM2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          CALM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CALM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CALM2 Antibody (H00000805-B01P) in WB

          Western Blot analysis of CALM2 expression in transfected 293T cell line by CALM2 MaxPab polyclonal antibody. Lane 1: CALM2 transfected lysate(16.5 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CALM2 Antibody in Western Blot (WB)

          Product Details

          H00000805-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CALM2 (AAH03354, 1 a.a. approximately 149 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKVKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAK

          Target Information

          Calmodulin 2 (CALM2) is one of the three human genes encoding calmodulin, a highly conserved calcium-binding messenger protein critical for mediating various cellular processes. Calmodulin proteins are composed of two pairs of EF-hand motifs located at both the N- and C-termini, connected by a flexible central linker. Each EF-hand motif binds calcium ions, with the C-terminus possessing two high-affinity Ca2+ binding sites, while the N-terminus contains two low-affinity sites. Upon binding calcium, calmodulin undergoes a conformational change that enables it to interact with and regulate numerous target proteins, including kinases, phosphatases, ion channels, and other enzymes. Specifically, calcium/calmodulin (CaM) signaling pathways are involved in critical cellular functions such as muscle contraction, neurotransmitter release, and cellular growth. The interaction of CALM2 with different targets is essential for translating transient changes in intracellular calcium levels into specific cellular responses, making it a pivotal player in calcium signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: calmodulin 2 (phosphorylase kinase, delta); Calmodulin-1; Calmodulin-2; Calmodulin-3; LP7057 protein; phosphorylase kinase delta; phosphorylase kinase subunit delta; prepro-calmodulin 2

          View more View less

          Gene Aliases: CALM; CALM1; CALM2; CALM3; CALML2; CAM; CAM1; CAM2; CAM3; CAMB; CAMC; CAMII; CAMIII; LQT15; PHKD; PHKD2

          View more View less

          UniProt ID: (Human) P0DP25

          View more View less

          Entrez Gene ID: (Human) 805

          View more View less

          Function(s)
          Ras guanyl-nucleotide exchange factor activity calcium ion binding protein binding adenylate cyclase binding inositol-1,4,5-trisphosphate 3-kinase activity ligand-gated ion channel activity protein kinase binding protein domain specific binding nitric-oxide synthase regulator activity titin binding type 3 metabotropic glutamate receptor binding thioesterase binding N-terminal myristoylation domain binding phospholipase binding protein serine/threonine kinase activator activity phosphatidylinositol 3-kinase binding ion channel binding calcium-dependent protein binding nitric-oxide synthase binding protein phosphatase activator activity
          Process(es)
          G2/M transition of mitotic cell cycle MAPK cascade response to amphetamine regulation of heart rate platelet degranulation detection of calcium ion glycogen catabolic process muscle contraction G-protein coupled receptor signaling pathway activation of adenylate cyclase activity Wnt signaling pathway, calcium modulating pathway positive regulation of peptidyl-threonine phosphorylation negative regulation of peptidyl-threonine phosphorylation regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion substantia nigra development regulation of rhodopsin mediated signaling pathway positive regulation of cyclic nucleotide metabolic process positive regulation of protein autophosphorylation regulation of cytokinesis positive regulation of phosphoprotein phosphatase activity ion transmembrane transport positive regulation of protein dephosphorylation Fc-epsilon receptor signaling pathway positive regulation of DNA binding positive regulation of GTPase activity inositol phosphate metabolic process regulation of nitric-oxide synthase activity positive regulation of nitric-oxide synthase activity positive regulation of cyclic-nucleotide phosphodiesterase activity response to corticosterone response to calcium ion regulation of cardiac muscle contraction negative regulation of ryanodine-sensitive calcium-release channel activity positive regulation of ryanodine-sensitive calcium-release channel activity positive regulation of protein serine/threonine kinase activity regulation of high voltage-gated calcium channel activity regulation of cell communication by electrical coupling involved in cardiac conduction
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0