Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGRKEEDDCS SWKKQTTNIR KTFIFMEVLG SGAFSEVFLV KQRLTGKLFA LKCIKKSPAF RDSSLENEIA VLKKIKHENI VTLEDIYEST THYYLVMQLV SGGELFDRIL ERGVYTEKDA SLVIQQVLSA VKYLHENGIV HRDLKPENLL YLTPEENSKI MITDFGLSKM EQNGIMSTAC GTPGYVAPEV LAQKPYSKAV DCWSIGVITY ILLCGYPPFY EETESKLFEK IKEGYYEFES PFWDDISESA KDFICHLLEK DPNERYTCEK ALSHPWIDGN TALHRDIYPS VSLQIQKNFA KSKWRQAFNA AAVVHHMRKL HMNLHSPGVR PEVENRPPET QASETSRPSS PEITITEAPV LDHSVALPAL TQLPCQHGRR PTAPGGRSLN CLVNGSLHIS SSLVPMHQGS LAAGPCGCCS SCLNIGSKGK SSYCSEPTLL KKANKKQNFK SEVMVPVKAS GSSHCRAGQT GVCLIM
CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase. Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK. CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium/calmodulin-dependent protein kinase IG; Calcium/calmodulin-dependent protein kinase type 1G; CaM kinase 1G; CaM kinase I gamma; CaM kinase IG; caM-KI gamma; CaMK-like CREB kinase III; caMKI gamma; caMKIG; CLICK III; RP1-272L16.2
Gene Aliases: CAMK1G; CLICK3; CLICKIII; dJ272L16.1; VWS1
UniProt ID: (Human) Q96NX5
Entrez Gene ID: (Human) 57172
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support