Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is predicted to react with bovine and rat based on sequence homology.
Potent and specific inhibitor of CaM-kinase II (CAMK2) (By similarity). Plays a role in the maintenance of long-term retrieval- induced memory in response to contextual fear (By similarity). Modulates blood pressure and vascular reactivity via regulation of CAMK2 activity in addition to regulation of left ventricular mass (By similarity). Mediates the NLRP3 inflammasome in cardiomyocytes via acting as an inhibitor of the MAPK14/p38 and MAPK8/JNK pathways, thereby regulating ventricular remodeling and cardiac rhythm post- myocardial infarction (By similarity). Negatively effects insulin sensitivity and promotes lipid formation in adipose tissues independent of CAMK2 signaling (By similarity).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcium/calmodulin-dependent protein kinase II inhibitor 1; calcium/calmodulin-dependent protein kinase II inhibitor alpha; CaMKII inhibitor protein alpha; CaMKII inhibitory protein alpha; CaMKIIN-alpha; CaMKIINalpha; mCaMKIINalpha; RP11-401M16.1
Gene Aliases: 1810006K23Rik; AI848599; AI891706; CAMK2N1; PRO1489
UniProt ID: (Human) Q7Z7J9, (Mouse) Q6QWF9
Entrez Gene ID: (Human) 55450, (Mouse) 66259
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support