Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CAMKK2 Antibodies

          Proteintech

          CAMKK2 Polyclonal Antibody

          32 References
          View all (10) CAMKK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CAMKK2 Polyclonal Antibody

          • Antibody Testing Data (9)
          CAMKK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          CAMKK2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CAMKK2 Antibody (11549-1-AP) in ICC/IF

          Immunofluorescent analysis of PC-3 cells using 11549-1-AP (CAMKK2 antibody) at dilution of 1:25 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CAMKK2 Antibody in Immunocytochemistry (ICC/IF)
          CAMKK2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CAMKK2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CAMKK2 Antibody in Western Blot (WB)
          CAMKK2 Antibody in Western Blot (WB)
          CAMKK2 Antibody in Western Blot (WB)
          CAMKK2 Antibody in Western Blot (WB)
          CAMKK2 Antibody in Western Blot (WB)
          CAMKK2 Antibody in Western Blot (WB)

          Product Details

          11549-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 25 publications 25 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          CAMKK2 Fusion Protein Ag2093 (1-353 aa encoded by BC026060)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.15 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSSCVSSQP SSNRAAPQDE LGGRGSSSSE SQKPCEALRG LSSLSIHLGM ESFIVVTECE PGCAVDLGLA RDRPLEADGQ EVPLDSSGSQ ARPHLSGRKL SLQERSQGGL AAGGSLDMNG RCICPSLPYS PVSSPQSSPR LPRRPTVESH HVSITGMQDC VQLNQYTLKD EIGKGSYGVV KLAYNENDNT YYAMKVLSKK KLIRQAGFPR RPPPRGTRPA PGGCIQPRGP IEQVYQEIAI LKKLDHPNVV KLVEVLDDPN EDHLYMVFEL VNQGPVMEVP TLKPLSEDQA RFYFQDLIKG IEYLHYQKII HRDIKPSNLL VGEDGHIKIA DFGVSNEFKG SDALLSNTVG TPAF (1-353 aa encoded by BC026060)

          Target Information

          CAMKK2 (CaMKK beta) is a serine/threonine kinase involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: calcium/calmodulin-dependent protein kinase beta; Calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; Calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; caM-kinase kinase beta; CaM-KK 2; CaM-KK beta; caMKK 2; caMKK beta; CAMKK beta protein; MGC15254

          View more View less

          Gene Aliases: 6330570N16Rik; AW061083; CAMKK; CAMKK2; CAMKKB; KIAA0787; mKIAA0787

          View more View less

          UniProt ID: (Human) Q96RR4, (Mouse) Q8C078

          View more View less

          Entrez Gene ID: (Human) 10645, (Mouse) 207565

          View more View less

          Function(s)
          calmodulin-dependent protein kinase activity protein tyrosine kinase activity calcium ion binding calmodulin binding ATP binding nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          MAPK cascade protein phosphorylation peptidyl-tyrosine phosphorylation calcium-mediated signaling regulation of protein kinase activity positive regulation of transcription, DNA-templated protein autophosphorylation phosphorylation activation of protein kinase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0