Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MANMQGLVER LERAVSRLES LSAESHRPPG NCGEVNGVIA GVAPSVEAFD KLMDSMVAEF LKNSRILAGD VETHAEMVHS AFQAQRAFLL MASQYQQPHE NDVAALLKPI SEKIQEIQTF RERNRGSNMF NHLSAVSESI PALGWIAVSP KPGPYVKEMN DAATFYTNRV LKDYKHSDLR HVDWVKSYLN IWSELQAYIK EHHTTGLTWS KTGPVASTVS AFSVLSSGPG LPPPPPPLPP PGPPPLFENE GKKEESSPSR SALFAQLNQG EAITKGLRHV TDDQKTYKNP SLRAQGGQTQ SPTKSHTPSP TSPKSYPSQK HAPVLELEGK KWRVEYQEDR NDLVISETEL KQVAYIFKCE KSTIQIKGKV NSIIIDNCKK LGLVFDNVVG IVEVINSQDI QIQVMGRVPT ISINKTEGCH IYLSEDALDC EIVSAKSSEM NILIPQDGDY REFPIPEQFK TAWDGSKLIT EPAEIMA
This gene was identified by its similarity to the gene for human adenylyl cyclase-associated protein. The function of the protein encoded by this gene is unknown. However, the protein appears to be able to interact with adenylyl cyclase-associated protein and actin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2810452G09Rik; Adenylyl cyclase-associated protein 2; CAP 2
Gene Aliases: CAP2
UniProt ID: (Human) P40123
Entrez Gene ID: (Human) 10486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support