Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Calpain 3 Antibodies

          Proteintech

          CAPN3 Monoclonal Antibody (1G12A5)

          View all (11) Calpain 3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CAPN3 Monoclonal Antibody (1G12A5)

          • Antibody Testing Data (3)
          CAPN3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          CAPN3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CAPN3 Antibody (67366-1-IG) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded mouse skeletal muscle tissue slide using 67366-1-Ig (CAPN3 antibody) at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CAPN3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CAPN3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          CAPN3 Antibody in Western Blot (WB)

          Product Details

          67366-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:3,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -
          Product Specifications

          Species Reactivity

          Mouse, Pig, Rat, Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1G12A5

          Immunogen

          CAPN3 Fusion Protein Ag13179

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1000 µg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Aliquoting is unnecessary for -20°C storage.

          Target Information

          Calpain, an intracellular calcium-dependent protease that cleaves cytoskeletal and submembranous proteins, plays a role in cytoskeletal reorganization and muscle protein degradation. Calpain is a heterodimer composed of a small regulatory subunit and one of three large catalytic subunits, designated calpain 1, calpain 2 and calpain 3. Calpain 3 (calpain p94) is a muscle-preferred calcium activated neutral protease that localizes to the nucleus. Mutations involving the calpain 3 gene are associated with limb-girdle muscle dystrophy type 2A, a form of autosomal recessive and progressive neuromuscular diso4. Calpastatin regulates calpain by inhibiting both the proteolytic activity of calpain and its binding to membranes. Calpastatin exists in two types, tissue type and erythrocyte type, resulting from both alternative splicing and proteolytic processing.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Calcium-activated neutral proteinase 3; calpain 3, (p94); Calpain L3; Calpain p94; calpain p94, large [catalytic] subunit; calpain, large polypeptide L3; Calpain-3; CANP 3; CAPN 3; LGMD 2A; MGC10767; MGC11121; MGC14344; MGC4403; Muscle-specific calcium-activated neutral protease 3; muscle-specific calcium-activated neutral protease 3 large subunit; nCL-1; New calpain 1; skeletal muscle specific calpain; skeletal muscle specific calpain p94

          View more View less

          Gene Aliases: AI323605; CANP3; CANPL3; Capa-3; Capa3; CAPN3; LGMD2; LGMD2A; Lp82; Lp84; Lp85; nCL-1; NCL1; p94

          View more View less

          UniProt ID: (Pig) P43368, (Human) P20807, (Mouse) Q64691, (Rat) P16259

          View more View less

          Entrez Gene ID: (Pig) 397349, (Human) 825, (Mouse) 12335, (Rat) 29155

          View more View less

          Function(s)
          catalytic activity calcium-dependent cysteine-type endopeptidase activity signal transducer activity calcium ion binding protein binding peptidase activity cysteine-type peptidase activity structural constituent of muscle sodium ion binding titin binding protein complex scaffold ligase regulator activity receptor binding hydrolase activity enzyme binding metal ion binding
          Process(es)
          autolysis protein complex assembly proteolysis positive regulation of satellite cell activation involved in skeletal muscle regeneration response to muscle activity myofibril assembly negative regulation of protein sumoylation negative regulation of apoptotic process regulation of I-kappaB kinase/NF-kappaB signaling sarcomere organization regulation of myoblast differentiation positive regulation of proteolysis negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated regulation of catalytic activity positive regulation of NF-kappaB transcription factor activity positive regulation of release of sequestered calcium ion into cytosol response to calcium ion muscle structure development G1 to G0 transition involved in cell differentiation cellular response to calcium ion cellular response to salt stress protein localization to membrane self proteolysis sodium-dependent self proteolysis response to drug apoptotic process signal transduction muscle organ development muscle cell cellular homeostasis negative regulation of skeletal muscle cell differentiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0