Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: YRKSYVRKAF MKLDFNKSGS VPIINIRKCY CAKKHSQVIS GHSTEEEIKS SFLETLKVAC SKSDEVSYGE FEDYYEGLSI GIVDDEDFVN ILRTPWGI
Calcyphosine-2 is a calcium-binding protein with 2 EF-hand motifs (Wang et al., 2002 [PubMed 11846421]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Calcyphosin-2; Calcyphosine-2; calcyphosphine 2
Gene Aliases: CAPS2; UG0636c06
UniProt ID: (Human) Q9BXY5
Entrez Gene ID: (Human) 84698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support