Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MADSSGQQGK GRRVQPQWSP PAGTQPCRLH LYNSLTRNKE VFIPQDGKKV TWYCCGPTVY DASHMGHARS YISFDILRRV LKDYFKFDVF YCMNITDIDD KIIKRARQNH LFEQYREKRP EAAQLLEDVQ AALKPFSVKL NETTDPDKKQ MLERIQHAVQ LATEPLEKAV QSRLTGEEVN SCVEVLLEEA KDLLSDWLDS TLGCDVTDNS IFSKLPKFWE GDFHRDMEAL NVLPPDVLTR VSEYVPEIVN FVQKIVDNGY GYVSNGSVYF DTAKFASSEK HSYGKLVPEA VGDQKALQEG EGDLSISADR LSEKRSPNDF ALWKASKPGE PSWPCPWGKG RPGWHIECSA MAG
This gene encodes a class 1 aminoacyl-tRNA synthetase, cysteinyl-tRNA synthetase. Each of the twenty aminoacyl-tRNA synthetases catalyzes the aminoacylation of a specific tRNA or tRNA isoaccepting family with the cognate amino acid. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alternative splicing of this gene results in multiple transcript variants encoding distinct isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CysRS; cysteine translase; cysteine tRNA ligase 1, cytoplasmic; Cysteine--tRNA ligase, cytoplasmic; Cysteinyl-tRNA synthetase; cysteinyl-tRNA synthetase, cytoplasmic
Gene Aliases: CA3; CARS; CARS1; CYSRS; MGC:11246
UniProt ID: (Human) P49589, (Mouse) Q9ER68
Entrez Gene ID: (Human) 833, (Mouse) 27267, (Rat) 293638
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support