Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPGSPVEVKI QSRSSPPTMP PLPPINPGGP RPVSFTPTAL SNGINHSPPT LNGAPSPPQR FSNGPASSTS SALTNQQLPA TCGARQLSKL KRFLTTLQQF GNDISPEIGE KVRTLVLALV NSTVTIEEFH CKLQEATNFP LRPFVIPFLK ANLPLLQREL LHCARAAKQT PSQYLAQHEH LLLNTSIASP ADSSELLMEV HGNGKRPSPE RREENSFDRD TIAPEPPAKR VCTISPAPRH SPALTVPLMN PGGQFHPTPP PLQHYTLEDI ATSHLYREPN KMLEHREVRD RHHSLGLNGG YQDELVDHRL TEREWADEWK HLDHALNCIM EMVEKTRRSM AVLRRCQESD REELNYWKRR YNENTELRKT GTELVSRQHS PGSADSLSND SQREFNSRPG TGYVPVEFWK KTEEAVNKVK IQAMSEVQKA VAEAEQKAFE VIATERARME QTIADVKRQA AEDAFLVINE QEESTENCWN CGRKASETCS GCNIARYCGS FCQHKDWERH HRLCGQNLHG QSPHGQGRPL LPVGRGSSAR SADCSVPSPA LDKTSATTSR SSTPASVTAI DTNGL
Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: core-binding factor, runt domain, alpha subunit 2; translocated to, 2; DKFZp313F2116; ETO homolog on chromosome 20; ETO homologous on chromosome 20; MTG8-like protein; MTG8-related protein 1; myeloid translocation gene-related protein 1; Myeloid translocation-related protein 1; p85; Protein CBFA2T2; RP5-1137F22.1; ZMYND3 CBFA2T2
Gene Aliases: CBFA2T2; EHT; MTGR1; p85; ZMYND3
UniProt ID: (Human) O43439
Entrez Gene ID: (Human) 9139
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support