Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MALAVAPWGR QWEEARALGR AVRMLQRLEE QCVDPRLSVS PPSLRDLLPR TAQLLREVAH SRRAAGGGGP GGPGGSGDFL LIYLANLEAK SRQVAALLPP RGRRSANDEL FRAGSRLRRQ LAKLAIIFSH MHAELHALFP GGKYCGHMYQ LTKAPAHTFW RESCGARCVL PWAEFESLLG TCHPVEPGCT ALALRTTIDL TCSGHVSIFE FDVFTRLFQP WPTLLKNWQL LAVNHPGYMA FLTYDEVQER LQACRDKPGS YIFRPSCTRL GQWAIGYVSS DGSILQTIPA NKPLSQVLLE GQKDGFYLYP DGKTHNPDLT ELGQAEPQQR IHVSEEQLQL YWAMDSTFEL CKICAESNKD VKIEPCGHLL CSCCLAAWQH SDSQTCPFCR CEIKGWEAVS IYQFHGQATA EDPGNSSDQE GRELELGQVP LSAPPLPPRP DLPPRKPRNA QPKVRLLKGN SPPAALGPQD PAPA
This gene encodes a member of the Cbl family of E3 ubiquitin ligases. Cbl proteins play important roles in cell signaling through the ubiquitination and subsequent downregulation of tyrosine kinases. Expression of this gene may be restricted to epithelial cells, and alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cas-Br-M (murine) ecotropic retroviral transforming sequence c; Cas-Br-M (murine) ectropic retroviral transforming sequence c; Cbl proto-oncogene C, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase C; E3 ubiquitin-protein ligase CBL-C; RING finger protein 57; RING-type E3 ubiquitin transferase CBL-C; SH3-binding protein CBL-3; SH3-binding protein CBL-C; Signal transduction protein CBL-C
Gene Aliases: CBL-3; CBL-SL; CBL3; CBLC; RNF57
UniProt ID: (Human) Q9ULV8
Entrez Gene ID: (Human) 23624
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support