Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CBLC Antibodies

          Abnova

          CBLC Monoclonal Antibody (3F10)

          View all (18) CBLC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CBLC Monoclonal Antibody (3F10)

          • Antibody Testing Data (1)
          CBLC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          CBLC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CBLC Antibody (H00023624-M01) in WB

          Western Blot analysis of CBLC expression in transfected 293T cell line by CBLC monoclonal antibody (M01), clone 3F10. Lane 1: CBLC transfected lysate (Predicted MW: 52.5 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CBLC Antibody in Western Blot (WB)

          Product Details

          H00023624-M01

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          3F10

          Immunogen

          CBLC (AAH28915, 1 a.a. approximately 474 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          Immunogen sequence: MALAVAPWGR QWEEARALGR AVRMLQRLEE QCVDPRLSVS PPSLRDLLPR TAQLLREVAH SRRAAGGGGP GGPGGSGDFL LIYLANLEAK SRQVAALLPP RGRRSANDEL FRAGSRLRRQ LAKLAIIFSH MHAELHALFP GGKYCGHMYQ LTKAPAHTFW RESCGARCVL PWAEFESLLG TCHPVEPGCT ALALRTTIDL TCSGHVSIFE FDVFTRLFQP WPTLLKNWQL LAVNHPGYMA FLTYDEVQER LQACRDKPGS YIFRPSCTRL GQWAIGYVSS DGSILQTIPA NKPLSQVLLE GQKDGFYLYP DGKTHNPDLT ELGQAEPQQR IHVSEEQLQL YWAMDSTFEL CKICAESNKD VKIEPCGHLL CSCCLAAWQH SDSQTCPFCR CEIKGWEAVS IYQFHGQATA EDPGNSSDQE GRELELGQVP LSAPPLPPRP DLPPRKPRNA QPKVRLLKGN SPPAALGPQD PAPA

          Target Information

          This gene encodes a member of the Cbl family of E3 ubiquitin ligases. Cbl proteins play important roles in cell signaling through the ubiquitination and subsequent downregulation of tyrosine kinases. Expression of this gene may be restricted to epithelial cells, and alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cas-Br-M (murine) ecotropic retroviral transforming sequence c; Cas-Br-M (murine) ectropic retroviral transforming sequence c; Cbl proto-oncogene C, E3 ubiquitin protein ligase; Cbl proto-oncogene, E3 ubiquitin protein ligase C; E3 ubiquitin-protein ligase CBL-C; RING finger protein 57; RING-type E3 ubiquitin transferase CBL-C; SH3-binding protein CBL-3; SH3-binding protein CBL-C; Signal transduction protein CBL-C

          View more View less

          Gene Aliases: CBL-3; CBL-SL; CBL3; CBLC; RNF57

          View more View less

          UniProt ID: (Human) Q9ULV8

          View more View less

          Entrez Gene ID: (Human) 23624

          View more View less

          Function(s)
          phosphotyrosine binding signal transducer activity epidermal growth factor receptor binding calcium ion binding zinc ion binding ligase activity SH3 domain binding ubiquitin protein ligase activity
          Process(es)
          cell surface receptor signaling pathway negative regulation of epidermal growth factor-activated receptor activity protein ubiquitination negative regulation of epidermal growth factor receptor signaling pathway protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of MAP kinase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0