Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Members of the precerebellin family, such as CBLN3, contain a cerebellin motif (see CBLN1; MIM 600432) and a C-terminal C1q signature domain (see MIM 120550) that mediates trimeric assembly of atypical collagen complexes. However, precerebellins do not contain a collagen motif, suggesting that they are not conventional components of the extracellular matrix (Pang et al., 2000 [PubMed 10964938]).[supplied by OMIM, Aug 2009].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cerebellin 3 precursor protein; Cerebellin-3; precerebellin 3
Gene Aliases: CBLN3; PRO1486; RGD1563731; UNQ755/PRO1486
UniProt ID: (Human) Q6UW01, (Mouse) Q9JHG0
Entrez Gene ID: (Human) 643866, (Mouse) 56410, (Rat) 501998
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support