FIGURE: 1 / 2
Store as concentrated solution.
Centrifuge briefly prior to opening vial.
CBP for CREB-binding protein is a 265 kDa nuclear protein which probably has many of the properties expected of a CREB co-activator. It shows substantial sequence homology with p300.