Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MEELSSVGEQ VFAAECILSK RLRKGKLEYL VKWRGWSSKH NSWEPEENIL DPRLLLAFQK KEHEKEVQNR KRGKRPRGRP RKLTAMSSCS RRSKLKVGGC AGYADPTSQH PLGVGGRQRE GLGPSGRGWH FCQQSVPLLG KQEPPFFLSL SFCCQGPQPA ESSSPPLPGA SCFSLSCTPL CWVAGSNCCR QALFPPRGSL GDGKEQEACV Q
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell division cycle associated 6; chromobox homolog 2 (Pc class homolog, Drosophila); Chromobox protein homolog 2; modifier 3; Pc class homolog
Gene Aliases: CBX2; CDCA6; M33; SRXY5
UniProt ID: (Human) Q14781
Entrez Gene ID: (Human) 84733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support