Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody recognizes CBX2 (Chromobox protein homolog 2). It binds to a folded domain, amino acids 2-70. CBX2 is a component of a Polycomb group (PcG) multiprotein PRC1-like complex, a complex class required to maintain the transcriptionally repressive state of many genes, including Hox genes, throughout development.
MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell division cycle associated 6; chromobox homolog 2 (Pc class homolog, Drosophila); Chromobox protein homolog 2; modifier 3; Pc class homolog
Gene Aliases: CBX2; CDCA6; M33; SRXY5
UniProt ID: (Human) Q14781
Entrez Gene ID: (Human) 84733
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support