Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MELSAVGERV FAAESIIKRR IRKGRIEYLV KWKGWAIKYS TWEPEENILD SRLIAAFEQK ERERELYGPK KRGPKPKTFL LKARAQAEAL RISDVHFSVK PSASASSPKL HSSAAVHRLK KDIRRCHRMS RRPLPRPDPQ GGSPGLRPPI SPFSETVRII NRKVKPREPK RNRIILNLKV IDKGAGGGGA GQGAGALARP KVPSRNRVIG KSKKFSESVL RTQIRHMKFG AFALYKPPPA PLVAPSPGKA EASAPGPGLL LAAPAAPYDA RSSGSSGCPS PTPQSSDPDD TLPKLLPETV SPSAPSWREP EVLDLSLPPE SAATSKRAPP EVTAAAGPAP PTAPEPAGAS SEPEAGDWRP EMSPCSNVVV TDVTSNLLTV TIKEFCNPED FEKVAAGVAG AAGGGGSIGA SK
Polycomb group (PcG) proteins form multiprotein complexes that regulate expression patterns of developmental and cell proliferation genes. CBX6 (chromobox protein homolog 6) is a 412 amino acid protein that is a component of the chromatin-associated PcG complex. Through interaction with PcG, CBX6 modifies chromatin and maintains the transcriptionally repressive state of genes. Members of the chromobox domain protein family are characterized by the presence of a 37 amino acid chromobox (CHRromatin Organization MOdifier) domain. This domain plays a mechanistic role in targeting chromodomain proteins to specific regions of the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: chromobox homolog 6; Chromobox protein homolog 6
Gene Aliases: CBX6
UniProt ID: (Human) O95503
Entrez Gene ID: (Human) 23466
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support