Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene belongs to the CFAP53 family. It was found to be differentially expressed by the ciliated cells of frog epidermis and in skin fibroblasts from human. Mutations in this gene are associated with visceral heterotaxy-6, which implicates this gene in determination of left-right asymmetric patterning. [provided by RefSeq, Aug 2015]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 53; Cilia- and flagella-associated protein 53; coiled-coil domain containing 11; Coiled-coil domain-containing protein 11
Gene Aliases: 4933415I03Rik; CCDC11; CFAP53; HTX6; RGD1306734
UniProt ID: (Human) Q96M91, (Mouse) Q9D439
Entrez Gene ID: (Human) 220136, (Mouse) 74453, (Rat) 364899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support