Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATADTPAPA SSGLSPKEEG ELEDGEISDD DNNSQIRSRS SSSSSGGGLL PYPRRRPPHS ARGGGSGGGG GSSSSSSSSQ QQLRNFSRSR HASERGHLRG PSSYRPKEPF RSHPPSVRMP SSSLSESSPR PSFWERSHLA LDRFRFRGRP YRGGSRWSRG RGVGERGGKP GCRPPLGGGA GSGFSSSQSW REPSPPRKSS KSFGRSPSRK QNYSSKNENC VEETFEDLLL KYKQIQLELE CINKDEKLAL SSKEENVQED PKTLNFEDQT STDNVSITKD SSKEVAPEEK TQVKTFQAFE LKPLRQKLTL PGDKNRLKKV KDGAKPLSLK SDTTDSSQGI PYRVKEGFTP IPGLKFSA
Subunit of the trimeric poly(A) tail exosome targeting (PAXT) complex, a complex that directs a subset of long and polyadenylated poly(A) RNAs for exosomal degradation. The RNA exosome is fundamental for the degradation of RNA in eukaryotic nuclei. Substrate targeting is facilitated by its cofactor MTREX, which links to RNA-binding protein adapters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 131; Coiled-coil domain-containing protein 131; MGC23401; proline/serine-rich coiled-coil 2; Proline/serine-rich coiled-coil protein 2; Zinc finger C3H1 domain-containing protein; zinc finger, C3H1-type containing
Gene Aliases: CCDC131; CSRC2; KIAA0546; PSRC2; ZFC3H1
UniProt ID: (Human) O60293
Entrez Gene ID: (Human) 196441
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support