Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASSASARTP AGKRVINQEE LRRLMKEKQR LSTSRKRIES PFAKYNRLGQ LSCALCNTPV KSELLWQTHV LGKQHREKVA ELKGAKEASQ GSSASSAPQS VKRKAPDADD QDVKRAKATL VPQVQPSTSA WTTNFDKIGK EFIRATPSKP SGLSLLPDYE DEEEEEEEEE GDGERKRGDA SKPLSDAQGK EHSVSSSREV TSSVLPNDFF STNPPKAPII PHSGSIEKAE IHEKVVERRE NTAEALPEGF FDDPEVDARV RKVDAPKDQM DKEWDEFQKA MRQVNTISEA IVAEEDEEGR LDRQIGEIDE QIECYRRVEK LRNRQDEIKN KLKEILTIKE LQKKEEENAD SDDEGELQDL LSQDWRVKGA LL
Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 16; Coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; orphan maintenance of genome 1; Zinc finger protein 830
Gene Aliases: CCDC16; OMCG1; ZNF830
UniProt ID: (Human) Q96NB3
Entrez Gene ID: (Human) 91603
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support