Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAEPGGAAGR SHPEDGSASE GEKEGNNESH MVSPPEKDDG QKGEEAVGST EHPEEVTTQA EAAIEEGEVE TEGEAAVEGE EEAVSYGDAE SEEEYYYTET SSPEGQISAA DTTYPYFSPP QELPGEEAYD SVSGEAGLQG FQQEATGPPE SRERRVTSPE PSHGVLGPSE QMGQVTSGPA VGRLTGSTEE PQGQVLPMGV QHRFRLSHGS DIESSDLEEF VSQEPVIPPG VPDAHPREGD LPVFQDQIQQ PSTEEGAMAE RVESEGSDEE AEDEGSQLVV LDPDHPLMVR FQAALKNYLN RQIEKLKLDL QELVVATKQS RAQRQELGVN LYEVQQHLVH LQKLLEKSHD RHAMASSERR QKEEELQAAR ALYTKTCAAA NEERKKLAAL QTEMENLALH LFYMQNIDQD MRDDIRVMTQ VVKKAETERI RAEIEKKKQG VGDTGAAFIP KTTQGDQVSG RGKKKPGTVV ARVSPAEL
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 40
Gene Aliases: CCDC40; CILD15; FAP172; KIAA1640
UniProt ID: (Human) Q4G0X9
Entrez Gene ID: (Human) 55036
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support