Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DELLNIAKS KQERTNSELH NLRQIYVKQQ SDLQFLNFNV ENSQELIQMY DSKMEESKAL DSSRDMCLSD LENNHPKVDI KREKNQKSLF KDQKFEAMLV QQNRSDKSSC DECKEKKQQI DTVFGEKSVI KLSSIFTKDL VEKHNLPWSL GGKTQIEPEN KITLCKIHTK SPKCHGTGVQ NEGKQPSETP TLSDEKQWHD VSVYLGLTNC PSSKHPEKLD VECQDQMERS EISCCQKNEA CLGESGMCDS KCCHPSNFII EAPGHMSDVE WMSIFKPSKM QRIVRLKSGC TCSESICGTQ HDSPASELIA IQDSHSLGSS KSALREDETE SSSNKKNSPT SLLIYKDAPA FNE (265-616 aa encoded by BC060796)
Nuclear receptor coactivator that can enhance preferentially estrogen receptors ESR1 and ESR2 transactivation. Modulates also progesterone/PGR, glucocorticoid/NR3C1 and androgen/AR receptors transactivation, although at lower level; little effect on vitamin D receptor/VDR. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: aaa-protein; cancer/testis antigen 109; coiled-coil domain-containing protein 60; Coiled-coil domain-containing protein 62; Protein aaa; Protein TSP-NY; testis-specific protein TSP-NY
Gene Aliases: AI661708; CCDC62; CT109; ERAP75; Gm150; TSP-NY
UniProt ID: (Human) Q6P9F0, (Mouse) E9PVD1
Entrez Gene ID: (Human) 84660, (Mouse) 208908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support