Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSERKVLNK YYPPDFDPSK IPKLKLPKDR QYVVRLMAPF NMRCKTCGEY IYKGKKFNAR KETVQNEVYL GLPIFRFYIK CTRCLAEITF KTDPENTDYT MEHGATRNFQ AEKLLEEEEK RVQKEREDEE LNNPMKVLEN RTKDSKLEME VLENLQELKD LNQRQAHVDF EAMLRQHRLS EEERRRQQQE EDEQETAALL EEARKRRLLE DSDSEDEAAP SPLQPALRPN PTAILDEAPK PKRKVEVWEQ SVGSLGSRPP LSRLVVVKKA KADPDCSNGQ PQAAPTPGAP QNRKEANPTP LTPGASSLSQ LGAYLDSDDS NGSN (1-323 aa encoded by BC000561)
Part of the spliceosome which catalyzes two sequential transesterification reactions, first the excision of the non-coding intron from pre-mRNA and then the ligation of the coding exons to form the mature mRNA (PubMed:29301961). Plays a role in stabilizing the structure of the spliceosome catalytic core and docking of the branch helix into the active site, producing 5'-exon and lariat intron-3'-intermediates. May protect cells from TP53-dependent apoptosis upon dsDNA break damage through association with PRP19-CD5L complex (PubMed:22952453). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Coiled-coil domain-containing protein 94; Splicing factor YJU2
Gene Aliases: 2900016D05Rik; AI413813; CCDC94; YJU2
UniProt ID: (Human) Q9BW85, (Mouse) Q9D6J3
Entrez Gene ID: (Human) 55702, (Rat) 501285, (Mouse) 72886
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support