Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This gene was identified by the interaction of its gene product with Grap2, a leukocyte-specific adaptor protein important for immune cell signaling. The protein encoded by this gene was shown to interact with cyclin D. Transfection of this gene in cells was reported to reduce the phosphorylation of Rb gene product by cyclin D-dependent protein kinase, and inhibit E2F1-mediated transcription activity. This protein was also found to interact with helix-loop-helix protein E12 and is thought to be a negative regulator of liver-specific gene expression. Two alternatively spliced variants, which encode distinct isoforms, have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cyclin D-type binding-protein 1; Cyclin-D1-binding protein 1; D-type cyclin-interacting protein 1; grap2 and cyclin D interacting protein; Grap2 and cyclin-D-interacting protein; Human homolog of Maid; Maternal Id-like protein; maternal inhibition of differentiation; SSEC-8; Stage specific embryonic cDNA-8 protein
Gene Aliases: AU022347; CCNDBP1; DIP1; GCIP; HHM; Maid; SECC-8; SSEC-8
UniProt ID: (Human) O95273, (Rat) Q5BK06, (Mouse) Q3TVC7
Entrez Gene ID: (Human) 23582, (Rat) 362201, (Mouse) 17151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support