Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD100 (SEMA4D) Antibodies

          Invitrogen

          CD100 Recombinant Human Monoclonal Antibody (5D12)

          View all (52) CD100 (SEMA4D) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite CD100 Recombinant Human Monoclonal Antibody (5D12)

          • Antibody Testing Data (3)
          CD100 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD100 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          CD100 Antibody (MA5-49983) in Flow

          Flow cytometry of CD. Samples were incubated with CD100 monoclonal antibody (Product # MA5-49983), washed and then followed by FITC-conjugated anti-Human IgG Fc antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          CD100 Antibody in Flow Cytometry (Flow)
          CD100 Antibody in Neutralization (Neu)
          CD100 Antibody in Neutralization (Neu)

          Product Details

          MA5-49983

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Neutralization (Neu)

          2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Human / IgG4

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5D12

          Immunogen

          Recombinant Human SEMA4D protein

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.03% Proclin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3093855

          Target Information

          Semaphorins are neuronal chemorepellants that direct pioneering neurons during nervous system development. CD100 (also known as semaphorin 4d), a 150 kDa homodimer, is a novel semaphorin that induces B cells to aggregate and improves their viability in vitro. CD100 modifies CD40-CD40L B cell signaling by augmenting B cell aggregation and survival and down regulating CD23 expression. CD72 is a lymphocyte receptor for the class IV semaphorin CD100, which is involved in regulating B cell signaling. CD100 is expressed on resting and PHA stimulated T cells. The protein is weakly expressed on NK cells, EBV transformed B cells, monocytes and tumor/peripheral blood T cell lines, and at higher density on activated T cells and B cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A8; BB18; CD100; GR3; M-Sema G; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 4D; sema domain, immunoglobulin domain (Ig), transmembrane domain (TM) and short cytoplasmic domain, 4D; Sema J; semaphorin H; Semaphorin-4D; Semaphorin-C-like 2; Semaphorin-J

          View more View less

          Gene Aliases: C9orf164; CD100; coll-4; M-sema-G; SEMA4D; Semacl2; SEMAJ; Semcl2

          View more View less

          UniProt ID: (Human) Q92854, (Mouse) O09126

          View more View less

          Entrez Gene ID: (Human) 10507, (Mouse) 20354

          View more View less

          Function(s)
          receptor activity transmembrane signaling receptor activity receptor binding protein binding semaphorin receptor binding neuropilin binding chemorepellent activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter neural crest cell migration positive regulation of protein phosphorylation immune response cell adhesion negative regulation of cell adhesion regulation of cell shape negative regulation of alkaline phosphatase activity positive regulation of phosphatidylinositol 3-kinase signaling positive regulation of cell migration regulation of cell projection organization negative regulation of apoptotic process positive regulation of GTPase activity ossification involved in bone maturation negative regulation of osteoblast differentiation positive regulation of collateral sprouting regulation of dendrite morphogenesis negative regulation of axon extension involved in axon guidance positive regulation of peptidyl-tyrosine phosphorylation negative regulation of peptidyl-tyrosine phosphorylation negative chemotaxis leukocyte aggregation semaphorin-plexin signaling pathway semaphorin-plexin signaling pathway involved in bone trabecula morphogenesis multicellular organism development nervous system development cell differentiation positive regulation of axonogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0